Hydroxylammoniumchlorid ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent (max. 0.01 ppm Hg)

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Hydroxylamine HCl, Hydroxylamine hydrochloride

24709.230EA 1610 DKK
24709.230
Hydroxylammoniumchlorid ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent (max. 0.01 ppm Hg)
Hydroxylammoniumchlorid
Formel: NH₂OH·HCl
Molekylvægt: 69.49 g/mol
Kogepunkt: 306 °C (1013 hPa)
Smeltepunkt: 151 °C
Massefylde: 1.67 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00051089
CAS-nummer: 5470-11-1
EINECS: 226-798-2
UN nummer: 2923
ADR: 8 (6.1) III
Merck Index: 13,04853

Bestil nu

Specifikationer Testresultater

Assay min. 99.0%
Solubility in alcohol passes test
Free acidity max 25 meq/g
pH (20°C; 5%) 2,5 - 3,5
Heavy metals (as Pb) max. 5 ppm
Ignition residue (SO4) max. 100 ppm
Insolubility in water max. 50 ppm
NH4 (Ammonium) max. 0.2%
SO4 (Sulphate) max. 20 ppm
As (Arsenic) max. 1 ppm
Cu (Copper) max. 10 ppm
Fe (Iron) max. 20 ppm
Hg (Mercury) max. 10 ppb
Pb (Lead) max . 5 ppm
Conforms to Reag.Ph.Eur. passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR