Formamid ≥99.0%, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Carbamaldehyde

24311.291EA 487 DKK
24311.291 24311.320
Formamid ≥99.0%, AnalaR NORMAPUR® analytical reagent
Formamid
Formel: HCONH₂
Molekylvægt: 45.04 g/mol
Kogepunkt: 210
Smeltepunkt: 2
Massefylde: 1.1334 g/cm³ (20 °C)
Flammepunkt: 79
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00007941
CAS-nummer: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay (on anhydrous substance) Min. 99.0 %
Heavy metals (as Pb) Max. 1 ppm
Ignition residue (SO4) Max. 50 ppm
Water Max. 0.2 %
Cl (Chloride) Max. 1 ppm
Fe (Iron) Max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR