Jern(III)sulfat-hydrat, GPR RECTAPUR®

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ferric Persulphate, Diiron trissulphate hydrate, Ferric sulphate hydrate

24248.292EA 1730 DKK
24248.292
Jern(III)sulfat-hydrat, GPR RECTAPUR®
Jern(III)sulfat-hydrat
Formel: Fe₂(SO₄)₃·xH₂O
Molekylvægt: 399.88 g/mol (anhy)
Massefylde: 3.1 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149714
CAS-nummer: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065

Bestil nu

Specifikationer Testresultater

Assay (calculated as Fe(III)) Min. 21 %
Cl (Chloride) Max. 100 ppm
Fe (II) (Iron) Max. 0.05 %
Zn (Zinc) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR