Sampling scoops, SteriPlast®
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5378-1007 5378-1001 5378-1002 5378-1003 5378-1004 5378-1005 5378-1006 5378-1008 5378-1013 5378-1014 5378-1015 5378-1016 5378-1019 5378-1020
231-0521EA 295 DKK
231-0521 231-0515 231-0516 231-0517 231-0518 231-0519 231-0520 231-0522 BURK5378-1013 BURK5378-1014 BURK5378-1015 BURK5378-1016 BURK5378-1019 331-0364
Sampling scoops, SteriPlast®
Øser
PS, white. These scoops are ideal for the sampling of powder, granules and pastes. They are also available with a transparent lid to close it immediately after the sampling procedure to secure a sterile and contamination-free sample.
  • Individually packed, gamma radiation sterilised
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines
Due to the special handle construction and the slightly angled edge the scoop stands horizontal and can be laid down without the material trickling out.