VWR®, Spatulas
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
231-0492EA 1670 DKK
231-0492
VWR®, Spatulas
Spatler
PS, white. This spatula, with its long, ergonomic solid handle and pointed blade, can easily pierce paper and plastic bags.
  • Production, assembling and packaging according to cleanroom Class 7 (10000)
  • According to EU food and FDA guidelines
  • Individually packed, sterilised by gamma radiation
Ideal for sampling powder, granules and pastes.