Pump tubing, Tygon® S3™ E-LFL
Leverandør: Saint Gobain Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AVX42003 AVX42007 AVX42012 AVX42022 AVX42017 AVX42019 AVX42029 AVX42038 AVX06057 070602-08-ND 070602-09-ND 070602-10-ND 070602-13-ND 070602-06-ND 070602-18-ND 070603-05-ND 070603-12-ND
228-1258EA 2740 DKK
228-1258 228-1259 228-1260 228-1262 228-1261 228-1321 228-1322 228-1323 228-1319 228-1329 228-1330 228-1331 228-1334 228-1327 228-1339 228-1343 228-1350
Pump tubing, Tygon® S3™ E-LFL
Slanger Overførselsslange
Non DEHP and phthalate-free tubing.
  • Over 1000 hours pump tubing life at 0 psi; over 600 hours pump tubing life at 10 psi
  • Extremely low particle spallation
  • Contains no BPA or phthalates
  • Broad chemical resistance
  • Meets USP Class VI, ISO 10993 and FDA criteria - suitable for pharma, medical and food applications
Longest flexible life of any clear Tygon® tubing. The non ageing and broad chemical resistance properties enable users to utilise the tubing for transferring chemicals, cleaning agents and bactericides without danger of corrosion (as with metals)More Product Information