Silikoneslanger, PharmaFocus®
Leverandør: Freudenberg Medical Europe GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
61-252-21 61-252-22 61-252-23 61-252-25 61-252-26 61-252-27 61-252-30 61-252-32 61-252-33 61-252-35 61-252-36
228-1168EA 944 DKK
228-1168 228-1169 228-1170 228-1171 228-1172 228-1173 228-1174 228-1175 228-1176 228-1177 228-1178
Silikoneslanger, PharmaFocus®
Slanger Overførselsslange
Tubing, silicone, platinum cured.
  • More cost-effective silicone tubing that delivers consistent quality and the highest level of assurance
  • Compliant with the requirements of USP, Class VI
  • FDA 21CFR 177-2600/ISO 10093-1/EP 3.1.9
The platinum-cured silicone elastomer used for the PharmaFocus tubing has been designed specifically to fit the needs of the pharmaceutical industry. This silicone undergoes a battery of biocompatibility tests appropriate for pharmaceutical applications, eliminating the costly extended implant studies typically performed onMore Product Information