Stopcocks, needle-valve, straight
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
02270002
227-3310EA 307 DKK
227-3310
Stopcocks, needle-valve, straight
Stophaner
Needle-valve stopcocks are made of DURAN® borosilicate glass, with PTFE spindle.
  • Screw cap and retention device made of PP
  • Temperature resistance from −20 to +150 °C
  • Excellent chemical resistance
These stopcocks are ideal for exact dispensing and therefore ideal for glass equipment.