Cerium (IV) sulfat tetrahydrat ≥98.0% analytical reagent

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Cerium disulphate tetrahydrate, Ceric sulphate tetrahydrate

22604.153EA 1370 DKK
22604.153
Cerium (IV) sulfat tetrahydrat ≥98.0% analytical reagent
Cerium (IV) sulfat tetrahydrat
Formel: Ce(SO₄)₂·4H₂O
Molekylvægt: 404.3 g/mol
Smeltepunkt: 180…200 °C
Massefylde: 3.91 g/cm³ (20 °C)
MDL Number: MFCD00149427
CAS-nummer: 10294-42-5
EINECS: 237-029-5
UN nummer: 3260
ADR: 8 III
Merck Index: 12,02036

Bestil nu

Specifikationer Testresultater

Assay Min. 98.0 %
Heavy metals (as Pb) Max. 50 ppm
Insolubility in diluted H2SO4 Max. 50 ppm
Cl (Chloride) Max. 10 ppm
PO4 (Phosphate) Max. 100 ppm
Fe (Iron) Max. 50 ppm

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR