Cadmiumsulfat 8/3-hydrat 98.0-102.0%, AnalaR NORMAPUR® ACS analytical reagent

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Cadmium sulphate, 8/3-hydrate, Cadmium sulphate octahydrate

22226.236EA 980 DKK
22226.236
Cadmiumsulfat 8/3-hydrat 98.0-102.0%, AnalaR NORMAPUR® ACS analytical reagent
Cadmiumsulfat 8/3-hydrat
Formel: 3CdSO₄·8H₂O
Molekylvægt: 256.51 g/mol (anhy)
Smeltepunkt: 41
Massefylde: 3.79 g/cm³ (20 °C)
MDL Number: MFCD02253000
CAS-nummer: 7790-84-3
EINECS: 233-331-6
UN nummer: 2570
ADR: 6.1 III
Merck Index: 13,01627

Bestil nu

Specifikationer Testresultater

Assay 98.0 - 102.0 %
Insoluble matter Max. 50 ppm
Cl (Chloride) Max. 10 ppm
NO3 + NO2 (as NO3) Max. 30 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 20 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 100 ppm
Na (Sodium) Max. 0.02 %
Pb (Lead) Max. 30 ppm
Zn (Zinc) Max. 0.05 %
Conforms to ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR