VWR®, Carboys, Narrow Neck, Round, with Screw Caps
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
216-1736EA 365 DKK
216-1736 216-1704 216-1703 216-1737 216-1706 216-1701 216-1729 216-1738 216-1702 216-1908 216-1918 216-2180
VWR®, Carboys, Narrow Neck, Round, with Screw Caps
Flasker Flasker, dunke
LDPE, HDPE or PP, translucent with purple screw caps. Use for storing and dispensing solutions and media. Round carboys feature an integrated shoulder handle. Graduated to identify the volume dispensed.
LDPE carboys
Ideal for high purity applications as they have high resistance to chemicals (widely used in ophthalmic product range, like eye droppers and nasal sprays).
Ideal resin for cold storage (down to −80 °C), working temperature −80 to +80 °C
Resistant to physical impact (will not break even if it falls accidentally, wheras PP carboys may lead to cracking at negative temperatures)
LDPEMore Product Information