Sølvnitrat 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Lunar caustic, Silver mononitrate, Nitric acid silver salt

21572.188EA 3690 DKK
21572.188 21572.235 21572.133 21572.292
Sølvnitrat 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Sølvnitrat
Formel: AgNO₃
Molekylvægt: 169.87 g/mol
Kogepunkt: 444
Smeltepunkt: 210
Massefylde: 4.35 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003414
CAS-nummer: 7761-88-8
EINECS: 231-853-9
UN nummer: 1493
ADR: 5.1 II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay 99.8 - 100.5 %
Clarity of solution Passes test ACS
Free acid Passes test ACS
Identification (Ag) Passes test Ph.Eur.
Identification (NO3) Passes test Ph.Eur.
Solution in water Passes test
Appearance of solution Passes test Ph.Eur.
Acidity or alkalinity Passes test Ph.Eur.
Aluminium, lead, copper and bismuth Passes test Ph.Eur.
Foreign salts ≤ 0.3 %
Insolubility in water ≤ 50 ppm
Not precipitated by HCl (as SO4) ≤ 100 ppm
Cl (Chloride) ≤ 5 ppm
SO4 (Sulphate) ≤ 20 ppm
Ca (Calcium) ≤ 10 ppm
Cd (Cadmium) ≤ 1 ppm
Cu (Copper) ≤ 2 ppm
Fe (Iron) ≤ 2 ppm
K (Potassium) ≤ 50 ppm
Mg (Magnesium) ≤ 5 ppm
Mn (Manganese) ≤ 5 ppm
Ni (Nickel) ≤ 5 ppm
Pb (Lead) ≤ 10 ppm
Zn (Zinc) ≤ 1 ppm
Conforms to Reag. Ph.Eur./ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR