Sølvnitrat ≥99.5%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Lunar caustic, Silver mononitrate, Nitric acid silver salt

21570.136EA 1280 DKK
21570.136 21570.182 21570.238 21570.295
Sølvnitrat ≥99.5%, GPR RECTAPUR®
Sølvnitrat
Formel: AgNO₃
Molekylvægt: 169.87 g/mol
Kogepunkt: 444
Smeltepunkt: 210
Massefylde: 4.35 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003414
CAS-nummer: 7761-88-8
EINECS: 231-853-9
UN nummer: 1493
ADR: 5.1 II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay Min. 99.5 %
Foreign heavy metals (as Pb) Max. 20 ppm
Not precipitated by HCl (as SO4) Max. 100 ppm
Cl (Chloride) Max. 10 ppm
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR