Dropper tips, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
242410 242401-01 242401 242404 242407 242424 242430 242431-01 242421 242421-01 242422 242423 242425 242426 242427 242428 242431 242418 242432 242433 242434 242436 242437 242438 242440 242402 242403 242405 242420 242408 242411 242411-01 242412 242413 242414 242415 242416 242417 242406 W242436-A W242431-A W242432-A W242433-A W242434-A W242437-A W242438-A W242440-A W242430-A W242401-01-A W242401-A W242402-A W242403-A W242404-A W242405-A W242406-A W242407-A W242408-A W242431-01-A W242410-A W242421-A W242422-A W242423-A W242424-A W242425-A W242420-A W242426-A W242427-A W242428-A W242415-A W242411-01-A W242411-A W242412-A W242413-A W242421-01-A W242416-A W242417-A W242418-A W242414-A
215-4784EA 401 DKK
215-4784 215-4150 215-4776 215-4779 215-4782 215-4797 215-4802 215-4153 215-4794 215-4152 215-4795 215-4796 215-4798 215-4799 215-4800 215-4801 215-4803 215-4792 215-4804 215-4805 215-4806 215-4807 215-4808 215-4809 215-4810 215-4777 215-4778 215-4780 215-4793 215-4783 215-4785 215-4151 215-4786 215-4787 215-4788 215-4789 215-4790 215-4791 215-4781 215-4835 215-4831 215-4832 215-4833 215-4834 215-4836 215-4837 215-4838 WHEAW242430-A 215-4811 215-4812 215-4813 215-4814 215-4815 215-4816 215-4817 215-4818 215-4819 215-4830 215-4820 WHEAW242421-A WHEAW242422-A WHEAW242423-A WHEAW242424-A WHEAW242425-A 215-4829 WHEAW242426-A WHEAW242427-A WHEAW242428-A 215-4826 215-4821 215-4822 215-4823 215-4824 WHEAW242421-01-A 215-4827 WHEAW242417-A 215-4828 215-4825
Dropper tips, WHEATON®
Låg Dråbetællerlåg
LDPE, coloured.
  • Extended controlled dropper tips
  • Dispense one drop at a time
  • Average drop size is 44 ±6 μl
Bestillingsoplysninger: Tips available in different colours and packaged in case of 100 or 1000.