Sterilin™ Quickstart Universal Containers, PS, 30 ml
Leverandør: STERILIN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
128C 128A 128B 128BBAC 128A/IRR 128AR/IRR
215-0324EA 1440 DKK
215-0324 215-0321 215-0322 215-0323 216-1556 216-1290
Sterilin™ Quickstart Universal Containers, PS, 30 ml
Prøvecontainere Allround kasser
Polystyrene, clear with QuickStart cap.
  • ¹/₂ - turn cap ensures improved end user handling
  • Suitable for centrifugation at 3800×g
  • Lot number on each container aids full traceability
  • Aseptically manufactured
  • In vitro use only
Secure excellent temperature and chemical resistance with these 95 kPa compliant 30 ml leak-resistant and CE marked Thermo Scientific™ Sterilin™ Quickstart Universal Polystyrene 30 ml containers. Improved performance, easy to handle. Available pre-filled with boric acid (0,4 g) for the preservation of urine samples.
Not availableMore Product Information