Test tubes, with rim, medium wall
Leverandør: MURRAY S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
G030/01 G030/02 G030/03 G030/04 G030/05 G030/06 G030/08 G030/10 G030/09
212-7097EA 638 DKK
212-7097 212-7098 212-7099 212-7100 212-7102 212-7103 212-0461 212-7104 MURRG030/09
Test tubes, with rim, medium wall
Rør Reagensglas
Borosilicate test tubes are suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples. They are also highly chemically resistant.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes