Test tubes without rim, medium wall
Leverandør: MURRAY S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
G002/83 G002/71 G002/73 G002/77 G002/81 G002/82 G002/84 G002/85 G002/86 G002/72 G002/76 G002/74 G002/75 G002/80
212-0559EA 1020 DKK
212-0559 212-7085 212-7087 212-7089 212-7090 212-7091 212-7092 212-7093 212-7094 212-7086 212-7088 212-0557 MURRG002/75 212-0558
Test tubes without rim, medium wall
Rør Reagensglas
Soda-lime glas, uden kant.
  • Good chemical and physical properties
  • Medium-walled
Suitable for use in applications that do not involve high thermal stresses, especially for bacteriological applications.