VWR®, Disposal Box for Pipette Tips
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
212-1833EA 1600 DKK
212-1833
VWR®, Disposal Box for Pipette Tips
Bokse Affaldsbeholdere
This acrylic box is ideal for the one-handed disposal of used pipette tips. The 10 mm thick acrylic provides protection from β radiation.
  • Quick flip of port cover makes tip ejection easy
  • Eliminates any need to handle pipette tips used with hazardous fluids
  • Large hinged lid allows easy disposal of contents
W×D×H: 154×171×206 mm