Ammoniumacetat 98,0 Soil_analysis

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Acetic acid ammonium salt

21199.361EA 2890 DKK
21199.361
Ammoniumacetat 98,0 Soil_analysis
Ammoniumacetat
Formel: H₃CCOONH₄
Molekylvægt: 77.08 g/mol
Smeltepunkt: 114
Massefylde: 1.17 g/cm³ (20 °C)
Flammepunkt: 136
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013066
CAS-nummer: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay Min. 98.0 %
pH (20°C; 5 %) 5.0 - 7.4
Heavy metals (as Pb) Max. 2 ppm
Ignition residue (SO4) Max. 50 ppm
Cl (Chloride) Max. 10 ppm
NO3 (Nitrate) Max. 10 ppm
SO4 (Sulphate) Max. 10 ppm
Ca (Calcium) Max. 5 ppm
Cu (Copper) Max. 0.5 ppm
Fe (Iron) Max. 2 ppm
K (Potassium) Max. 1 ppm
Mg (Magnesium) Max. 1 ppm
Na (Sodium) Max. 1 ppm
Pb (Lead) Max. 0.5 ppm
Zn (Zinc) Max. 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR