Ammoniumjern(III)sulfat dodecahydrat ≥98%, TECHNICAL

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Alum iron, Ammonium iron sulphate dodecahydrate, Ferric ammonium sulphate dodecahydrate, Ammonium iron(III) sulphate dodecahydrate, Iron alum, Ammonium ferric sulphate dodecahydrate

21092.293EA 378 DKK
21092.293
Ammoniumjern(III)sulfat dodecahydrat ≥98%, TECHNICAL
Ammoniumjern(III)sulfat dodecahydrat
Formel: NH₄Fe(SO₄)₂·12H₂O
Molekylvægt: 482.2 g/mol
Kogepunkt: 230
Smeltepunkt: 39…41 °C
Massefylde: 1.71 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00150004
CAS-nummer: 7783-83-7
EINECS: 233-382-4
Merck Index: 12,00549

Bestil nu

Specifikationer Testresultater

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR