Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent

Leverandør: VWR Chemicals
Ratings:
Gennemsntilig vurdering: 3.7 (Bedømmelse)

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethyl alcohol

20821.296EA 284 DKK
20821.296 20821.310 20821.321 20821.365 20821.467 20821.558 20821.330 VWRC20821.558DP 20821.930
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Ethanol

Advarsel: WARNING: This alcohol is regulated. Please contact VWR.

Formel: C₂H₅OH
Molekylvægt: 46.07 g/mol
Kogepunkt: 77.5…78.5 °C
Smeltepunkt: –117 °C
Massefylde: 0.7895 g/cm³ (20 °C)
Flammepunkt: 12 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003568
CAS-nummer: 64-17-5
EINECS: 200-578-6
UN nummer: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Bestil nu

Specifikationer Testresultater

Assay (on anhydrous substance) Min. 99.8 %
Assay (V/V) Min. 99.85 %
IR Spectrum Passes test
Solubility in water Passes test ACS
Volatile impurities Passes test Ph.Eur.
Acidity Max. 0.0001 meq/g
Alkalinity Max. 0.0001 meq/g
Boiling point 77.5 - 78.5 °C
Colouration Max. 10 APHA
Density (20/20) 0.790 - 0.793
Refractive index (20°C) 1.360 - 1.362
Substances coloured by H2SO4 Max. 20 APHA
Aldehydes (as CH3CHO) Max. 5 ppm
Aldehydes + ketones (as CH3CHO) Max. 10 ppm
iso-Amyl alcohol Max. 0.05 %
Benzene Max. 2 ppm
Evaporation residue Max. 5 ppm
Formaldehyde Max. 1 ppm
Furaldehyde Max. 1 ppm
Higher alcohols (standard mixture) Max. 0.02 %
Ketones (as CH3COCH3) Max. 10 ppm
Methanol Max. 100 ppm
2-Propanol Max. 30 ppm
Substances reducing KMnO4 (as O) Max. 3 ppm
Total heavy alcohols Max. 100 ppm
Water Max. 0.1 %
Al (Aluminium) Max. 0.1 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.2 ppm
Cd (Cadmium) Max. 0.02 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.01 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 0.05 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.05 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm
Absorbance (240 nm) (5 cm) Max. 0.4
Absorbance (250-260 nm) (5 cm) Max. 0.3
Absorbance (270-340 nm) (5 cm) Max. 0.1
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR