Kaliumthiocyanat 99,0 ACS, Fluka™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Potassium rhodanide

207799-100G 207799-2.5KG
207799-100GEA 251 DKK
207799-100G 207799-2.5KG
Kaliumthiocyanat 99,0 ACS, Fluka™
Potassium thiocyanate
Formel: KSCN
Molekylvægt: 97.18 g/mol
Kogepunkt: 500 Dec. (on heating)
Smeltepunkt: 172 °C
Massefylde: 1.89 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011413
CAS-nummer: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Bestil nu

Specifikationer Testresultater

Assay Min. 98.5 %
Water insoluble matter Max. 0.005 %
pH (5 %, 25 °C) 5.3 - 8.7
Ammonium (NH₄) Max. 3 ppm
Iron (Fe) Max. 2 ppm
Sodium (Na) Max. 0.005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.005 %
Matter consuming I (as I) complying
IR spectrum complying

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR