Dimethylarsinsyre natriumsalt, Millipore®

Leverandør: Merck Millipore Calbiochem
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

205541-100
205541-100EA 4360 DKK
205541-100
Dimethylarsinsyre natriumsalt, Millipore®
Dimethylarsinsyre natriumsalt
Formel: C₂H₆AsNaO₂
Molekylvægt: 159.94 g/mol
CAS-nummer: 124-65-2
EINECS: 204-708-2
UN nummer: 1688
ADR: 6.1 II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR