EDTA 99 zA

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II

20301.186EA 300 DKK
20301.186 20301.290
EDTA 99 zA
EDTA
Formel: C₁₀H₁₆N₂O₈
Molekylvægt: 292.25 g/mol
Kogepunkt: 614
Smeltepunkt: 220
Massefylde: 0.86 g/cm³ (20 °C)
Flammepunkt: >100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003541
CAS-nummer: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay Min. 99 %
Heavy metals (as Pb) Max. 20 ppm
Ignition residue (SO4) Max. 0.2 %
Water Max. 0.2 %
Cl (Chloride) Max. 0.05 %
SO4 (Sulphate) Max. 0.05 %
Fe (Iron) Max. 10 ppm

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR