Anti-IgG Goat Polyclonal Antibody (CF™ 594)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20160 20160-1
20160.EA 2240 DKK
20160. 20160-1
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
Antistoffer
Type: Secondary
Antigen: IgG (Pig) (pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR