Cæsiumchlorid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Cesium Chloride

2012.0100EA 0 DKK
2012.0100
Cæsiumchlorid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Cæsiumchlorid
Formel: CsCl
Molekylvægt: 168.36 g/mol
Kogepunkt: 1303
Smeltepunkt: 646
Massefylde: 3.7808 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010955
CAS-nummer: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021

Bestil nu

Specifikationer Testresultater

Assay min. 99.5 %
Nitrogen Compounds (as N) max. 0.001 %
Trace Impurities (in ppm)
Aluminium (Al) max. 5
Iron (Fe) max. 3
Potassium (K) max. 20
Magnesium (Mg) max. 5
Sodium (Na) max. 20
Heavy Metals (as Pb) max. 5
Sulfate (SO4) max. 50


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR