Filtration funnels, semi-micro, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SF4T31
201-8686EA 1090 DKK
201-8686
Filtration funnels, semi-micro, Quickfit®
Tragte Filtreringstragte Tragte med konisk slib
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for filtering semi-micro quantities of material.