Multiple adapters, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MA4/23
201-3002EA 1050 DKK
201-3002
Multiple adapters, Quickfit®
Destillations komponenter Forskellige Adaptere
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With three necks. Two vertical and one angled at 45°.