Ammoniumsulfit monohydrat 92

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

17996.A1 17996.A7
17996.A1EA 673 DKK
17996.A1 17996.A7
Ammoniumsulfit monohydrat 92
Ammoniumsulfit monohydrat

Purity: 92% min CAS No.: 7783-11-1 Molecular Formula: H8N2O3S.H2O Formula Weight: 134.15 (116.14anhy) Physical Form: Crystalline Density: 1.41 Refractive Index: 1.5150 Sensitivity: Air Sensitive Appearance: White Melting Point: 60-70° dec. Merck Reference: 14,557 MDL No.: MFCD00066094

Formel: H₈N₂O₃S·1H₂O
Molekylvægt: 134.16 g/mol
Smeltepunkt: 60…70 °C (dec.)
Massefylde: 1.41
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00066094
CAS-nummer: 7783-11-1
EINECS: 233-484-9

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR