Supelco® Organic Reference Standard, Triglyceride Mix
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17810-1AMP-S
17810-1AMP-SEA 358 DKK
17810-1AMP-S
Supelco® Organic Reference Standard, Triglyceride Mix
Standarder Økologiske standarder
Emballering: 100 mg ampule contains approximately equal amounts by weight of: Triacetin (C2:0) Tributyrin (C4:0) Tricaproin (C6:0) Tricaprylin (C8:0) Tricaprin (C10:0)