Methanol 99.9% for analysis

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

176840010 176840025 176845000 176840250 176840100
176840010.EA 342 DKK
176840010. 176840025. 176845000. 176840250. 176840100.
Methanol 99.9% for analysis
Methanol
Formel: CH₃OH
Molekylvægt: 32.04 g/mol
Kogepunkt: 64.7 °C (760 mmHg)
Smeltepunkt: –98 °C
Massefylde: 0.791 g/cm³
Flammepunkt: 12 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004595
CAS-nummer: 67-56-1
EINECS: 200-659-6
UN nummer: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear liquid
Water ≤0.05 % (K.F.)
Solubility (in water) Passes test
GC ≥99.85 %
Color scale ≤5 APHA
Residue after evaporation ≤0.001 %
Acidity/alkalinity ≤0.0005 meq/g
Substance darkened by H2SO4 ≤5 APHA
Calcium (Ca) ≤0.2 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Potassium (K) ≤0.5 ppm
Sodium (Na) ≤0.5 ppm
Zinc (Zn) ≤0.05 ppm
Carbonyl compounds ≤0.001 %
Ethanol ≤0.05 %
2-Propanol ≤0.005 %
KMnO4-reducing substance ≤0.00025 %
Total phosphorus (P) ≤0.05 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR