VWR®, Ultrapure Water System, G Series
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
171-1094EA 123090 DKK
171-1094 171-1096 171-1095 171-1464 171-1465 171-1466
VWR®, Ultrapure Water System, G Series
Vandrensningssystemer Rene og ultrarene vandsystemer
This powerful, intuitive laboratory water purification system provides superior and consistent ultrapure Type I and EDI-based Type II water from tap water. Water quality meets or exceeds ASTM, CLSI, CAP and ISO Type I water standards, respectively.
  • Simple and intuitive to dispense water via the dispenser or central console
  • Central console is an 8 inch waterproof touch screen, providing user with full control and access of system performance
  • One water system can drive several dispensing units ('1 + N mode')
  • Dispensing flow rate is up to 2 L/min and