Ultrapure water purification systems, Milli-Q® EQ 7000
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EQ7000HMI SMEQ00KT WMEQ0DKT WMEQ0RKT BTEQ0DKT
171-1399EA 11730 DKK
171-1399 171-1429 171-1430 171-1431 171-1478
Ultrapure water purification systems, Milli-Q® EQ 7000
Vandrensningssystemer Rene og ultrarene vandsystemer
To deliver reliable, high quality scientific results, you require consistently high quality ultrapure water. The Milli-Q® EQ 7000 ultrapure water system is designed to produce consistent ultrapure water quality [resistivity 18,2 MΩ·cm @ 25 °C; total organic carbon (TOC) ≤5 ppb] that can be adapted to each user’s specific application requirements. Plus, a range of intelligent design features make it effortless to dispense water as needed and where needed in the lab. An ultrapure water system designed by scientists, for scientists.
  • Three flow rates or one-touch volumetric dispensing