(2-Hydroxyethyl)trimethylammoniumchlorid 98.0-100.5%, BAKER ANALYZED®, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-hydroxy-N,N,N-trimethylethanaminium chloride

1582-01EA 642 DKK
1582-01
(2-Hydroxyethyl)trimethylammoniumchlorid 98.0-100.5%, BAKER ANALYZED®, J.T.Baker®
Choline chloride

Lot analysis on label.

Formel: HOH₂CCH₂N(CH₃)₃Cl
Molekylvægt: 139.62 g/mol
Smeltepunkt: 244…247 °C
Massefylde: ∼1.205 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011721
CAS-nummer: 67-48-1
EINECS: 200-655-4
Merck Index: 13,02226
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b015e76f45084bc46da3249578a85652cd6802&vid=85c8a0a1b152b861b3bc3ec8fe25a70509221d85&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

For Laboratory,Research,or Manufacturing Use 98.0 - 100.5 %
Assay (anhydrous basis) 98.0 - 100.5 %
Residue after Ignition ≤0.10 %
Ammonium Compounds Passes Test
Heavy Metals (as Pb) ≤0.003 %
Iron (Fe) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR