Natriumchlorat ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

14267.22 14267.A1 14267.D9
14267.22EA 442 DKK
14267.22 14267.A1 14267.D9
Natriumchlorat ≥99%
Sodium chlorate

Purity: 99% min CAS No.: 7775-09-9 Molecular Formula: NaClO3 Formula Weight: 106.45 Physical Form: Granular Density: 2.5 Melting Point: 248° Solubility: Soluble in water Merck Reference: 14,8598 UN No.: 1495 MDL No.: MFCD00003479

Formel: ClNaO₃
Molekylvægt: 106.44 g/mol
Kogepunkt: 300
Smeltepunkt: 248 °C
Massefylde: 2.5
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003479
CAS-nummer: 7775-09-9
EINECS: 231-887-4
UN nummer: 1495
ADR: 5.1 II
REACH: 01-2119474389-23
Merck Index: 13,08670

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR