Natriumnitrit ≥97% ACS

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Diazotizing salts

14244.14 14244.22 14244.36
14244.14EA 297 DKK
14244.14 14244.22 14244.36
Natriumnitrit ≥97% ACS
Sodium nitrite

Purity: 97% min CAS No.: 7632-00-0 Molecular Formula: NaNO2 Formula Weight: 69.00 Physical Form: Granular Density: 2.17 Melting Point: 271° Solubility: Soluble in water Merck Reference: 14,8648 UN No.: 1500 MDL No.: MFCD00011118

Formel: NaNO₂
Molekylvægt: 69 g/mol
Kogepunkt: 320
Smeltepunkt: 271 °C
Massefylde: 2.17
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011118
CAS-nummer: 7632-00-0
EINECS: 231-555-9
UN nummer: 1500
ADR: 5.1 III
Merck Index: 14,8648

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR