Lead standard dissolved in oil, 1,000 µg/g Pb in mineral oil Specpure® for wear metal analysis

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

14208.18
14208.18EA 1180 DKK
14208.18
Lead standard dissolved in oil, 1,000 µg/g Pb in mineral oil Specpure® for wear metal analysis
Standarder Kationiske standarder Lead Standards

Physical form: liquid
MDL No.: MFCD00134050

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR