Production and laboratory chairs, with permanent contact backrest, Neon 2
Leverandør: bimos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9563-MG01-3278 9563-SP02-3280 9563-2000-3278 9563-2000-3279 9563-2000-3280 9563-2001-3278 9563-2001-3280 9563-MG01-3279 9563-MG01-3280 9563-MG02-3278 9563-MG02-3279 9563-MG02-3280 9563-SP01-3278 9563-SP01-3279 9563-SP01-3280 9563-SP02-3278 9563-2001-3279 9563-SP02-3279 9563-2002-3278 9563-2002-3279 9563-2002-3280 9563-MG11-3279 9563-MG11-3280 9563-MG11-3278 9563-SP11-3278 9563-SP11-3279 9563-SP11-3280 9563-6801-3280 9563-6801-3279 9563-6801-3278 9563-6802-3278 9563-6802-3279 9563-6802-3280 9563-6811-3278 9563-6811-3279 9563-6811-3280 9563-2000-3277 9563-6801-3277 9563-MG01-3277 9563E-98113280 9563E-MG013280 9563E-MG013278 9563E20003278 9563-SP01-3277 9563MG01327852 9563E-98013278 9563-SP02-3277 9563E20003279 9563EMG013279 9563E20003277 9563E20003280
132-0913EA 6910 DKK
132-0913 139-0657 132-0910 132-0911 132-0912 139-0631 139-0633 132-0914 132-0915 139-0643 139-0644 139-0645 139-0649 139-0653 139-0654 139-0655 139-0632 139-0656 139-0634 139-0635 139-0636 139-0647 139-0648 139-0646 139-0658 139-0659 139-0660 139-0437 132-0917 132-0916 139-0637 139-0638 139-0639 139-0640 139-0641 139-0642 132-0979 132-0981 132-0980 BIMO9563E-98113280 132-1416 132-1415 132-1414 BIMO9563-SP01-3277 132-1417 BIMO9563E-98013278 BIMO9563-SP02-3277 BIMO9563E20003279 BIMO9563EMG013279 BIMO9563E20003277 BIMO9563E20003280
Production and laboratory chairs, with permanent contact backrest, Neon 2
Stole
Neon supports the forward leaning sitting position that is often necessary in lab work. Its modern design matches the high-tech laboratory environment perfectly and has all the characteristics you would expect of a good laboratory chair. It is seamless, easy to clean, washable, and resistant to disinfectants making Neon the ideal solution for the laboratory when a really comfortable ergonomic chair is needed.
  • Castors
  • Permanent contact backrest
  • Ergonomic package with height, seat inclination, seat depth and backrest height adjustment and, weight regulation