SMS sequential sterilisation wraps, blue, heavy duty, 50 gsm
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KCRP-300-01.5 KCRP-300-0202 KCRP-300-0303 KCRP-300-0404 KCRP-300-0606 KCRP-300-0808 KCRP-300-0909 KCRP-300-0912 KCRP-300-1212 KCRP-300-1515 KCRP-300-1818 KCRP-300-1822 KCRP-300-2020 KCRP-300-2424 KCRP-300-3030 KCRP-300-3636 KCRP-300-4040 KCRP-300-4545 KCRP-300-4548 KCRP-300-4848 KCRP-300-5454 KCRP-300-5472 KCRP-300-6060 KCRP-300-6080
129-3751EA 3710 DKK
129-3751 129-3752 129-3753 129-3754 129-3755 129-3756 129-3757 129-3758 129-3759 129-3760 129-3761 129-3762 129-3763 129-3764 129-3765 129-3766 129-3767 129-3768 129-3769 129-3770 129-3771 129-3772 129-3773 129-3774
SMS sequential sterilisation wraps, blue, heavy duty, 50 gsm
Steriliseringsindpakning
Keystone Cleanroom Products' line of heavy-duty SMS wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
  • Manufactured and packaged in a certified ISO 8 cleanroom
  • Clean - no cellulose
  • Strong, rip and tear resistant 50 gsm SMS material
  • An extensive range of sizes available means less cutting
  • Hydrophobic barrier allows sterilisation gases, but prevents liquid and microbial penetration
  • Sub packed into sealed labelled poly bags with lot number and date of manufacture for complete traceability
For use in steam,More Product Information