Autoclavable Peel Pouches, with Steam Indicator, Heat Seal, 1073B Tyvek® / Polyester
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BBMGTVKHS0306 BBMGTVKHS0415 BBMGTVKHS0608 BBMGTVKHS0610 BBMGTVKHS0612 BBMGTVKHS0810 BBMGTVKHS0815 BBMGTVKHS1020 BBMGTVKHS1030 BBMGTVKHS1212 BBMGTVKHS1217 BBMGTVKHS1218 BBMGTVKHS1224 BBMGTVKHS1617 BBMGTVKHS1822 BBMGTVKHS2418 BBMGTVKHS2424 BBMGTVKHS2430 BBTMGTVK1840 BBTMGTVK2440 BBTMGTVK2442 BBTMGTVK2448 BBMGTVKHS0206 BBMGTVKHS0208 BBMGTVKHS02095 BBMGTVKHS0308 BBMGTVKHS03508 BBMGTVKHS0422 BBMGTVKHS0510 BBMGTVKHS0620 BBMGTVKHS0625 BBMGTVKHS0628 BBMGTVKHS0632 BBMGTVKHS0635 BBMGTVKHS0710 BBMGTVKHS0713 BBMGTVKHS0807 BBMGTVKHS0814 BBMGTVKHS0818 BBMGTVKHS0834 BBMGTVKHS1011 BBMGTVKHS1012 BBMGTVKHS1015 BBMGTVKHS1214 BBMGTVKHS1220 BBMGTVKHS1420 BBMGTVKHS1424 BBMGTVKHS1516 BBMGTVKHS1520 BBMGTVKHS1616 BBMGTVKHS1618 BBMGTVKHS1620 BBMGTVKHS1624 BBMGTVKHS1626 BBMGTVKHS1630 BBMGTVKHS1632 BBMGTVKHS1635 BBMGTVKHS1816 BBMGTVKHS2426 BBMGTVKHS2435
129-3354EA 9110 DKK
129-3354 129-3355 129-3356 129-3357 129-3358 129-3359 129-3292 129-3293 129-3294 129-3295 129-3296 129-3297 129-3298 129-3299 129-3300 129-3301 129-3302 129-3303 129-3367 129-3369 129-3370 129-3371 KACCBBMGTVKHS0206 KACCBBMGTVKHS0208 KACCBBMGTVKHS02095 KACCBBMGTVKHS0308 KACCBBMGTVKHS03508 KACCBBMGTVKHS0422 KACCBBMGTVKHS0510 129-3748 KACCBBMGTVKHS0625 KACCBBMGTVKHS0628 KACCBBMGTVKHS0632 KACCBBMGTVKHS0635 KACCBBMGTVKHS0710 KACCBBMGTVKHS0713 KACCBBMGTVKHS0807 KACCBBMGTVKHS0814 KACCBBMGTVKHS0818 KACCBBMGTVKHS0834 KACCBBMGTVKHS1011 129-3749 KACCBBMGTVKHS1015 KACCBBMGTVKHS1214 KACCBBMGTVKHS1220 KACCBBMGTVKHS1420 KACCBBMGTVKHS1424 KACCBBMGTVKHS1516 KACCBBMGTVKHS1520 KACCBBMGTVKHS1616 KACCBBMGTVKHS1618 KACCBBMGTVKHS1620 KACCBBMGTVKHS1624 KACCBBMGTVKHS1626 KACCBBMGTVKHS1630 129-3750 KACCBBMGTVKHS1635 KACCBBMGTVKHS1816 KACCBBMGTVKHS2426 KACCBBMGTVKHS2435
Autoclavable Peel Pouches, with Steam Indicator, Heat Seal, 1073B Tyvek® / Polyester
Poser Steriliseringsposer
Improve the performance, microbial barrier and cleanliness of your autoclave peel pouches.
  • Manufactured and packed in a controlled cleanroom environment
  • Integrated steam sterilisation indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • One side made of hydrophobic and lint-free Tyvek® 1073B
  • One side made of laminated polyester/polyolefin film
  • Peels open cleanly
  • Heat seal
A significant upgrade over paper-based steam sterilisation pouches. These Tyvek® peel pouches will not rip or puncture like paper-based autoclave pouches routinelyMore Product Information