Metal-in static shielding bags, SCS
Leverandør: DESCO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1001012 1001216 1001818 1001824 100810 100812 10035 10046 10058 100610 10068 3001012 3001014 3001418 3001824 30035 30046 30058 300610 30068 300810 1001024 1001224 3001024
129-2609EA 224 DKK
129-2609 129-2610 129-2611 129-2612 129-2613 129-2614 129-2622 129-2623 129-2624 129-2625 129-2626 129-2631 129-2632 129-2633 129-2634 129-2635 129-2636 129-2637 129-2638 129-2639 129-2640 DESC1001024 DESC1001224 DESC3001024
Metal-in static shielding bags, SCS
Poser Opbevaringsposer
Transparent metal-in static shielding bag provides a static safe environment for ESD sensitive electronic devices (ESDS).
  • Printed with an ESD protective symbol and a lot code for traceability
  • Heat sealable or zip-locked
  • Manufactured from a polyester, metal, polyethylene laminate
The polyester dielectric in concert with the metal layer provides discharge shielding. The exterior being static dissipative allows electrostatic charges to be removed when grounded.
Certificeringer: Complies with IEC 61340-5-1 and IEC 61340-5-3.
Emballering: Suppied perMore Product Information