Disposal containers, Allpack Manyside
Leverandør: Allpack
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HPE40
129-1300EA 0 DKK
129-1300
Disposal containers, Allpack Manyside
Bokse Affaldsbeholdere
Cardboard box, PE bag.
  • Bag is fully protected by Serflex, which makes the container tamper-proof
  • Easy assembly
  • Non toxic during incineration as no toxic pollutants are released
  • Stackable for storage
The Allpack Manyside high security box is an octagonal, reinforced corrugated cardboard container containing a PE bag welded at several points. Two external transport handles avoid contact with the bag. Double opening: A flip cover that can be left open to create a large opening, and a smaller opening.
Oval slot (mm): 270×100