Bags, Hazard BinLiner™
Leverandør: MICRONOVA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BL2-25REIRBB BL2-32YEIRBB BL4-32YEIRBB BH2-24YEIRBB
129-1088EA 2460 DKK
129-1088 129-1597 129-1589 129-1653
Bags, Hazard BinLiner™
Poser Affaldsposer
These low density polyethylene Hazard BinLiner™ bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.
  • Low density polyethylene
  • Hazardous material bags are labelled for waste segregation in the U.S.
  • Smaller bags with a star bottom are ideal for round containers
  • Larger bags are flat sealed
  • Can be used with BLSSH series BinLiner™ Stands
  • Good compatibility with common cleaners
  • Gamma irradiated liners are sterile for aseptic areas
The Biohazard liners are designed for containment and disposal ofMore Product Information