Rørformet posefilm til sterilisering
Leverandør: S▄DPACK MEDICA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7600547 7600513 7600515 7600518 7600519 7600523 7600526 7600532 7600538 7600546 7600567 7600508
129-1086EA 5460 DKK
129-1086 129-1422 129-1423 129-1434 129-1425 129-1426 129-1435 129-1411 129-1412 129-1430 129-1437 129-0485
Rørformet posefilm til sterilisering
Poser Steriliseringsposer
Tubular bag film is available as a flat or gussetted bag in various sizes. Helpful tips for use are printed on the film. Suitable for autoclaving and ethylene oxide sterilisation.
* Gusset's dimension.