Strontiumoxid, tech.

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

12495.30 12495.A1 12495.A7
12495.30EA 1520 DKK
12495.30 12495.A1 12495.A7
Strontiumoxid, tech.
Strontium oxide

CAS No.: 1314-11-0 Molecular Formula: SrO, may also contain Sr(OH)2, SrCO3 Formula Weight: 103.62 Physical Form: -100 Mesh Powder Density: 4.700 Sensitivity: Moisture Sensitive Boiling Point: ca 3000° Solubility: Soluble in fused potassium hydroxide. Insoluble in ether and acetone. Decomposes in water to Sr(OH)2 with evolution of heat Merck Reference: 14,8846 UN No.: 3262 MDL No.: MFCD00049558


In the manufacture of strontium salts, pyrotechnics, and pigments, in greases and soaps, and as a desiccant

Formel: SrO
Kogepunkt: ∼3000 °C
Smeltepunkt: 2430 °C
Massefylde: 4,700
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00049558
CAS-nummer: 1314-11-0
EINECS: 215-219-9
UN nummer: 3262
ADR: 8,II
Merck Index: 14,8846

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR