Acid red 1, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Red 2G, C.I. 18050, Amido Naphthol Red G, Sodium 5-acetamido-4-hydroxy-3-(phenyldiazenyl)naphthalene-2,7-disulphonate, 5-Acetamido-4-hydroxy-3-phenylazo-2,7-naphthalene-disulphonic acid disodium salt, Pontacyl Carmine, Azophloxine

11640-25G
11640-25GEA 314 DKK
11640-25G
Acid red 1, Sigma-Aldrich®
Acid red 1

Azophloxine has been used as a dye in Von Kossa staining and nonlinear optical studies. It is also used in photocatalytic tests.

Formel: C₁₈H₁₅N₃O₈S₂
Molekylvægt: 509.41 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003954
CAS-nummer: 3734-67-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR