Toilet cleaners, SURE®
Leverandør: Diversey
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
100891589 100891611
115-3072EA 341 DKK
115-3072 115-3073
Toilet cleaners, SURE®
Renseopløsninger Disinfektanter
Ready to use, heavy-duty cleaner for effective removal of lime scale and other deposits from toilet bowls and urinals.
  • Quickly dissolves calcium/lime deposits
  • Long lasting action on vertical/sloped surfaces
  • Easily reaches hidden areas such as under the toilet rim
  • Cleans without leaving residue
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product in use for user
SURE® toilet cleaner is part of a unique range of plant-based,More Product Information