EBP bar
Leverandør: DESCO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
231275
115-3044EA 106 DKK
115-3044
EBP bar
Jordforbindelser Jordledninger
The EBP bar should be fixed to the edge of a bench or other suitable work equipment within reach and accessible to all connecting ground cords, but where it is unlikely to be knocked and damaged or hinder the operator.
  • Mount EBP bar with two M5 pan-head screws (included), minimum 12 mm long
  • Connect earthing cord to a suitable earth. A spare ring terminal is supplied if the cordneeds to be shortened
The bar is 72×37×38 mm in size and is made of ABS. The yellow bar has arear cover to prevent access to the resistors.
The bars are available with aMore Product Information