Sterile delta wipes, PROSAT®
Leverandør: CONTEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PS-7030IR-BPR
115-2739EA 8520 DKK
115-2739
Sterile delta wipes, PROSAT®
Papirservietter Servietter til Renrum Imprægneret
100% nonwoven polyester wipes presaturated with 70% IPA and 30% DI water.
  • Suitable for Grade A/B or ISO Class 5-8 environments
  • Hydroentangled polyester fabric is highly sorbent with good wet strength
  • Low in particles and fibres
  • Nonscratching and ideal for general cleaning applications where a sterile wipe is required
  • Presaturated to reduce solvent usage and VOC emissions
PROSAT® Sterile Delta Wipes are made from a 100% nonwoven polyester fibres which are hydroentangled together using no binders or additives. The resulting fabric is extremely soft,More Product Information