Servietter
Leverandør: Zellstoff-Vertriebs-GmbH & Co. KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11338-01 11324-01 11160 11165 11490
115-2720EA 933 DKK
115-2720 115-2721 115-2722 115-2723 115-2724
Servietter
Papirservietter Genanvendelige rengøringsklude Klude til alment brug
Multitex® strong, reusable: Particularly tear-resistant viscose fleece wipes, suitable for wet and dry cleaning. The cloths are solvent-resistant and they meet food safety requirements. Multitex® cloths can be washed out after cleaning and reused.
Polytex®, wet-use: Chemical resistant wipes made of 100% polypropylene. Washable and reusable. These tear-proof, fibre-free wipes are a clean and economic solution for many applications.
Multisoft® Cell: White, 4 ply construction, soft and with very high absorptive capacity. For cleaning and polishing applications.
Multiclean® Plus: HighlyMore Product Information